Europium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11300.06 11300.14 11300.22
11300.06EA 130 EUR
11300.06 11300.14 11300.22
Europium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Europium(III) oxide

Purity: 99.9% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: Powder Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997

Formule: Eu₂O₃
Smeltpunt: 2291 °C
Dichtheid: 7,42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010997
CAS nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR