Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

11304.14 11304.22
11304.14EA 109 EUR
11304.14 11304.22
Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium trichloride hydrate

Purity: 99.9% CAS No.: 19423-85-9 Molecular Formula: ErCl3.xH2O Formula Weight: 273.62(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00010989

Formule: Cl₃Er·H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010989
CAS nummer: 19423-85-9
EINECS: 233-385-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR