Cadmium ≥99.99% (metals basis), powder -200 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11353.06 11353.14 11353.22
11353.06EA 90.5 EUR
11353.06 11353.14 11353.22
Cadmium ≥99.99% (metals basis), powder -200 mesh
Cadmium

Purity: 99.99% CAS No.: 7440-43-9 Appearance: Silvery white UN No.: 3179 MDL No.: MFCD00010914

Formule: Cd
MW: 112.41 g/mol
Kookpunt: 765
Smeltpunt: 321
Dichtheid: 8.64 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010914
CAS nummer: 7440-43-9
EINECS: 231-152-8
UN: 3179
ADR: 4.1 II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR