Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aluminum

11373.FI 11373.GH 11373.HP
11373.FIEA 73.6 EUR
11373.FI 11373.GH 11373.HP
Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Aluminium

Purity: 99.998% CAS No.: 7429-90-5 Physical Form: ≈6.75g/50x50mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029

Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR