Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11386.FIEA 150 EUR
11386.FI 11386.GH
Indium ≥99.9975% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Indium

Purity: 99.9975% CAS No.: 7440-74-6 Physical Form: ≈1.8g/50x50mm Odour: Odourless MDL No.: MFCD00134048

Formule: In
MW: 114.82 g/mol
Kookpunt: 2000
Smeltpunt: 156
Dichtheid: 7.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR