Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11392.FIEA 157 EUR
11392.FI 11392.GH
Copper ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Copper

Purity: 99.9985% CAS No.: 7440-50-8 Physical Form: ≈11.2g/50x50mm Odour: Odourless MDL No.: MFCD00010965

Formule: Cu
Kookpunt: 2595 °C (1013 hPa)
Smeltpunt: 1083 °C
Dichtheid: 8,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR