ESD safety shoes, slip-on, JALAS® 3520
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3520-36 3520-37 3520-38 3520-39 3520-40 3520-41 3520-42 3520-43 3520-44 3520-45 3520-46 3520-47
114-7889EA 121.43 EUR
114-7889 114-7890 114-7891 114-7892 114-7893 114-7894 114-7895 114-7896 114-7897 114-7898 114-7899 114-7900
ESD safety shoes, slip-on, JALAS® 3520
Schoenen en laarzen
Anti-static ESD safety shoes made from water repellent, PU coated leather upper plus a PU, oil resistant outsole.
  • Double shock-absorption zones
  • Exchangeable textile EVA insoles
  • Orthopaedic insoles possible
  • Toe protection cap made from aluminium
Colour: White
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2 SRC
Order Now
Specificaties
Environmentally Preferable