Shoe covers, PP/CPE
Leverancier: Hubei Orient International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
114-9036EA 177 EUR
114-9036 114-9045 114-9046 HOIT110179 114-9040 114-9041 HOIT110490 114-9038 114-9039 HOIT40042M-XL-BULK HOIT110017 114-9035
Shoe covers, PP/CPE
Wegwerpkleding
These non woven shoe covers are made of PP with a skid-resistant CPE coating on the sole.
  • High degree of sensitivity and high strength
  • Latex-free
  • Elasticated opening
Ideal for use in hospitals and medical environments.
Weight: 38 g/m² (PP fabric)
Order Now
Specificaties
Environmentally Preferable
Geslacht
Kledingstuk toepassing Medical,Clinical