Tychem® 6000 AL TF630 WG Ventilated Coveralls
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TF0630TGYWG/3X TF0630TGYWG/L TF0630TGYWG/M TF0630TGYWG/S TF0630TGYWG/XL TF0630TGYWG/2X
114-9614EA 526 EUR
114-9614 114-9611 114-9610 114-9609 114-9612 114-9613
Tychem® 6000 AL TF630 WG Ventilated Coveralls
Wegwerpkleding
Air-Fed coverall with attached non-dssipative undergloves and integrated dissipative socks.
  • Ventilated coverall with stitched and overtaped seams
  • Fully enclosed over hood with clear front and crown see-through visor
  • Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
  • Three exhaust valves; two at hips, one in hood back
  • Air distribution made of Tychem 6000 fabric
  • Emergency egress
  • Attached non dissipative undergloves
  • Integrated dissipative socks
Tychem® 6000 AL garments are made from a lightweight and durableMore Product Information