Silver ≥99.997% (metals basis), draad, Premion®, Ø 0.1 mm (0.004 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11470.G2 11470.G6 11470.H2
11470.G2EA 58.4 EUR
11470.G2 11470.G6 11470.H2
Silver ≥99.997% (metals basis), draad, Premion®, Ø 0.1 mm (0.004 in)
Silver

Purity: 99.997% CAS No.: 7440-22-4 Physical Form: ≈0.08g/m Odour: Characteristic MDL No.: MFCD00003397

Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Smeltpunt: 962 °C
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR