Cleanroom doekjes, ProWipe® 750
Leverancier: BERKSHIRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PW750121220 PW750.0909.20 PW750.090920U
115-0351EA 49.1 EUR
115-0351 115-0536 BERKPW750.090920U
Cleanroom doekjes, ProWipe® 750
Schoonmaakdoekjes Cleanroom doekjes Droge cleanroom doekjes
Non woven polypropylene/cellulose blend, for general applications where visible fibre contamination is a concern. Wipes have durable fibre construction for tough cleaning and are useful for spill control and wet areas.
  • Multi-layer composite with cellulose layer thermally bonded between the two polypropylene outer layers
  • Engineered for compatibility with acid solvents
  • Extremely durable and pure with very low particle and fiber generation
  • High absorbency to weight ratio may reduce cost in use compared to similar weight nonwovens
  • Chemically compatible with
More Product Information