Cleanroom anti-fatigue mats, Softline
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SL0206 SL0203 SL0220 SL0420 SL0209 SL0412 SL0406 SL0320 SL0306 SL0304 SL-GR0420 SL-BLU0320 SL-BLU0312 SL-BLU0309 SL-BLU0404 SL-BLU0406 SL-BLU0409 SL-BLU0209 SL-BLU0212 SL-BLU0412 SL-BLU0220 SL-BLU0303 SL-BLU0304 SL-BLU0306 SL-GR0220 SL-GR0304 SL-GR0212 SL-GR0306 SL-GR0309 SL-GR0303 SL-GR0204 SL-GR0203 SL-GR0206 SL-GR0209 SL-BLU0420 SL0212 SL0204 SL0409 SL0404 SL-BLU0203 SL-BLU0204 SL-BLU0206 SL0312 SL0303 SL0309 SL-GR0312 SL-GR0320 SL-GR0404 SL-GR0406 SL-GR0409 SL-GR0412
115-0796EA 358 EUR
115-0796 115-0795 115-0798 115-0804 115-0797 115-0803 115-0802 115-0801 115-0800 115-0799 ERGOSL-GR0420 ERGOSL-BLU0320 ERGOSL-BLU0312 ERGOSL-BLU0309 ERGOSL-BLU0404 ERGOSL-BLU0406 ERGOSL-BLU0409 ERGOSL-BLU0209 ERGOSL-BLU0212 ERGOSL-BLU0412 ERGOSL-BLU0220 ERGOSL-BLU0303 ERGOSL-BLU0304 ERGOSL-BLU0306 ERGOSL-GR0220 ERGOSL-GR0304 ERGOSL-GR0212 ERGOSL-GR0306 ERGOSL-GR0309 ERGOSL-GR0303 ERGOSL-GR0204 ERGOSL-GR0203 ERGOSL-GR0206 ERGOSL-GR0209 ERGOSL-BLU0420 ERGOSL0212 ERGOSL0204 ERGOSL0409 ERGOSL0404 ERGOSL-BLU0203 ERGOSL-BLU0204 ERGOSL-BLU0206 ERGOSL0312 ERGOSL0303 ERGOSL0309 ERGOSL-GR0312 ERGOSL-GR0320 ERGOSL-GR0404 ERGOSL-GR0406 ERGOSL-GR0409 ERGOSL-GR0412
Cleanroom anti-fatigue mats, Softline
Vloermatten Anti-vermoeidheidsmatten
The Ergomat Softline is made from nitrile rubber and is designed to increase comfort while reducing tripping hazards, body aches, quality issues, and sick leaves. It is designed for areas where resistance to oil, petrochemicals and heat is important. Ideal for wet environments, in case of excessive volumes of fluids, the Softline can be die-cut to allow for drainage.
  • Upper side with taps
  • Autoclavable, withstands 600 °C (welding)
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevent
More Product Information