Cleanroom doekjes, B'Wipes 6000
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
115-1821EA 88.6 EUR
115-1821 115-1823
Cleanroom doekjes, B'Wipes 6000
Schoonmaakdoekjes Cleanroom doekjes Droge cleanroom doekjes
2 ply polyester sealed-border wipe for use in ISO 3 cleanrooms.
  • Quilted polyester wipe, constructed from 2 plies, with sealed borders
  • Laundered, with sealed borders, for excellent cleanliness
  • 2 ply quilted, for extreme absorption
  • Ideal for spill control in critical areas, and for applying a cleaning/disinfecting solution over a large surface