Cleanroom NovaSponges
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP4-533 SP68-46 SP68-745 SP20-46 SP6-363 SP68-46IR SPAA-363 SP6-745 SP2-46 SP6-33 SP68-33 SP6-46
129-0639EA 654 EUR
129-0639 129-0641 115-1877 129-0638 MICRSP6-363 MICRSP68-46IR 115-1815 MICRSP6-745 MICRSP2-46 115-1817 MICRSP68-33 MICRSP6-46
Cleanroom NovaSponges
Schoonmaakdoekjes Sponzen
Cleanroom sponges to meet every demand from isolators and laminar flow benches to equipment interiors and laboratory applications. Choose from a range of fabric covers for specialized cleaning.
  • Made with cleanroom approved materials
  • Durable, reusable and absorbent
  • Fabric covers securely sewn over reticulated urethane foam interiors
  • Shape and size options available
  • Available gamma irradiated for aseptic areas
  • Hold up to strong disinfectants
  • Autoclavable
NovaCel™ Sponge: Ultra-clean polyvinyl acetate, firm to the touch, but soft and pliant once wetted -More Product Information