ESD floor mattings, Notrax® 457 Skystep™
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
457S0023BL 457S0034BL 457S0035BL
115-2864EA 538 EUR
115-2864 115-2865 115-2866
ESD floor mattings, Notrax® 457 Skystep™
Vloermatten Anti-vermoeidheidsmatten
The Skystep™ ESD static dissipative mat is a stand-alone rubber mat designed for individual work stations. Ergonomically engineered, the SkyStep™ ESD utilizes the power of air.
  • Superb anti-fatigue properties due to the ergonomic bubble pattern
  • Rubber molded, air-pocket underside anti-fatigue mat
  • Made of heavy duty natural rubber compound
  • ESD Static Dissipative, measured resistance 105 to 107 Ω
  • Beveled edges on all four sides
  • Grounding cord available
The mat is designed to suction to the ground to prevent shifting while trapping air in its chambers.More Product Information