PIG® Grippy® adhesive-backed absorbent mat
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MAT3250
115-2942EA 185 EUR
115-2942
PIG® Grippy® adhesive-backed absorbent mat
Absorptiematerialen en bescherming tegen morsen
This exclusive, adhesive-backed mat stays put to prevent slips, trips and falls around workstations and walkways near machinery in industrial workplaces.
  • Adhesive-backed absorbent mat stays exactly where you put it with no shifting, bunching up or flipping over to prevent slips, trips and falls in workstations and around machinery
  • Proprietary adhesive sticks to the floor with a super-tight grip, but peels up easily for quick change outs; reduces clean-up time to make your workplace more productive
  • PIG Grippy mat will stick to most common commercial floor
More Product Information