Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11509.FF 11509.FI 11509.GH
11509.FFEA 451 EUR
11509.FF 11509.FI 11509.GH
Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
Platinum

Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈1.34g/25x25mm Odour: Odourless MDL No.: MFCD00011179

Formule: Pt
Kookpunt: 3827 °C (1013 hPa)
Smeltpunt: 1769 °C
Dichtheid: 21,447 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR