Titanium(IV) fluoride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11540.14 11540.22
11540.14EA 121 EUR
11540.14 11540.22
Titanium(IV) fluoride ≥98%
Titanium(IV) fluoride

Purity: 98% CAS No.: 7783-63-3 Molecular Formula: TiF4 Formula Weight: 123.89 Physical Form: Crystalline Density: 2.798 Appearance: White Odour: Odourless Melting Point: 284° subl. Merck Reference: 14,9479 UN No.: 3260 MDL No.: MFCD00042545

Formule: F₄Ti
Kookpunt: 284 °C (1013 hPa)
Dichtheid: 2,798
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042545
CAS nummer: 7783-63-3
EINECS: 232-017-6
UN: 3260
ADR: 8,II
Merck Index: 14,9479

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR