Potassium chloride 99.0-100.5% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

11595.A1 11595.A7 11595.30
11595.A1EA 74.2 EUR
11595.A1 11595.A7 11595.30
Potassium chloride 99.0-100.5% ACS
Potassium chloride

Purity: 99.0-100.5% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: Crystalline Density: 1.984 Refractive Index: 1.490 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 773° Boiling Point: 1500° subl. Solubility: Freely soluble in water. Soluble in glycerol, alcohol Merck Reference: 14,7621 MDL No.: MFCD00011360


In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy

Formule: KCl
Kookpunt: 1500 °C (sublimatie)
Smeltpunt: 773 °C
Dichtheid: 1,984
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,7621

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR