Diammonium hydrogen orthophosphate ≥98.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

11597.30 11597.A1
11597.30EA 38 EUR
11597.30 11597.A1
Diammonium hydrogen orthophosphate ≥98.0% ACS
Diammonium hydrogen orthophosphate

Purity: 98.0% min CAS No.: 7783-28-0 Molecular Formula: (NH4)2HPO4 Formula Weight: 132.06 Physical Form: Crystalline Density: 1.619 Appearance: White Melting Point: 155° dec. Merck Reference: 14,542 MDL No.: MFCD00010891

Formule: (NH₄)₂HPO₄
Dichtheid: 1,619
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010891
CAS nummer: 7783-28-0
EINECS: 231-987-8
Bereik: 01-2119490974-22
Merck Index: 14,542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR