Autoclave indicator tape
Leverancier: Synergy Health
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31002550
116-0426EA 0 EUR
116-0426
Autoclave indicator tape
Sterilisatie-indicatoren
Crepe paper coated with a dry natural rubber based adhesive for steam sterilisation.
  • Good adhesion on wrap made with cellulose fibres
  • Good resistance to sterilisation process
  • Indicator colour change: from green to brown/black
Products must be stored in their original packaging, in dry and cool areas away from humidity.
It’s recommended to use the tape within two years after manufacturing.
Certificaten: Conformity ISO 11140-1, CE Mark.