Prohibition signs
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
822443 822447 822001 822150 823040 822147 823189 823190 824679 824680 822444 822445 824381 824382 825424 825425 822295 822296 823785 823041 825722 825723 831090 831091 822891 822892 821848 821849 823786 824828 824829 824977 824978 824530 824531 821997 821998 822146 822746 824231 823337 223654 803588 803586
BRDY822443EA 3.28 EUR
BRDY822443 BRDY822447 BRDY822001 BRDY822150 116-0464 116-0439 116-0440 116-0441 116-0442 116-0443 116-0444 116-0445 116-0446 116-0447 116-0448 116-0449 116-0452 116-0453 116-0456 116-0465 116-0466 116-0467 116-0468 116-0469 116-0470 116-0471 116-0472 116-0473 116-0457 116-0458 116-0459 116-0460 116-0461 116-0462 116-0463 116-0436 116-0437 116-0438 BRDY822746 BRDY824231 BRDY823337 BRDY223654 BRDY803588 BRDY803586
Prohibition signs
Pictogrammen
Prohibition signs must have a black diagram encircled by a red band and dissected by a red diagonal bar.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm
  • Different diameters
Order Now
Specificaties
Environmentally Preferable