Standard hazard warning signs
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
826616 828687 829723 828096 827800 816667 816668 827948 827949 827504 827505 827356 827357 829132 829133 828097 830612 830613 829428 829429 828688 828689 828836 828837 827801 829724 829725 829872 829873 830464 830465 830020 830021 826617 830463 829421 828685 829721 830609
116-0506EA 3.97 EUR
116-0506 BRDY828687 116-0686 116-0484 116-0494 116-0474 116-0475 116-0476 116-0477 116-0478 116-0479 116-0480 116-0481 116-0482 116-0483 116-0485 116-0486 116-0487 116-0488 116-0489 116-0490 116-0491 116-0492 116-0493 116-0495 116-0496 116-0497 116-0498 116-0499 116-0500 116-0501 116-0502 116-0503 116-0507 BRDY830463 BRDY829421 BRDY828685 BRDY829721 BRDY830609
Standard hazard warning signs
Pictogrammen
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm