Lithium perchlorate, anhydrous ≥95.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11639.09 11639.22 11639.36
11639.09EA 36.5 EUR
11639.09 11639.22 11639.36
Lithium perchlorate, anhydrous ≥95.0% ACS
Lithium perchlorate

Purity: 95.0% min CAS No.: 7791-03-9 Molecular Formula: ClLiO4 Formula Weight: 106.40 Physical Form: Crystalline Density: 2.43 Appearance: White Melting Point: 236° Boiling Point: 400° dec. Solubility: Partially soluble in water (30% at 25°, 72% at 100°). Soluble in alcohol, acetone, ether, ethyl acetate Merck Reference: 14,5539 UN No.: 1481 MDL No.: MFCD00011079


As an oxidizing agent

Formule: ClLiO₄
MW: 106.39 g/mol
Kookpunt: 400 °C (ontbinding)
Smeltpunt: 236 °C
Dichtheid: 2.43
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011079
CAS nummer: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1 II
Merck Index: 14,5539

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR