Eyewash station, HypaClens
Leverancier: SAFETY FIRST AID GROUP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E458
118-0283EA 59.9 EUR
118-0283
Eyewash station, HypaClens
Eerste-hulpbenodigdheden Oogdouches
This wall-mountable eyewash station holds two 500 ml HypaClens eyewash bottles.
  • Integrated mirror allows for self-administration of eyewash
  • Compact design of the eyewash station makes it ideal for any work place with low risks
W×D×H (mm): 250×60×300
Bestelinformatie: Station is supplied with two bottles of eyewash solution. Wall fixings are not included. Refills are available separately, please contact your local Avantor sales office for further information.