Cadmiumchloride, anhydrous ≥99.99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cadmium chloride

11860.14 11860.22
11860.14EA 126 EUR
11860.14 11860.22
Cadmiumchloride, anhydrous ≥99.99% (metals basis)
Cadmiumchloride

Purity: 99.99% CAS No.: 10108-64-2 Molecular Formula: CdCl2 Formula Weight: 183.32 Physical Form: Powder Density: 4.047 Melting Point: 568° Boiling Point: 960° Merck Reference: 14,1617 UN No.: 2570 MDL No.: MFCD00010916

Formule: CdCl₂
MW: 183.32 g/mol
Kookpunt: 960 °C
Smeltpunt: 568 °C
Dichtheid: 4.047
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010916
CAS nummer: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1 III
Merck Index: 14,1617

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR