Cadmium ≥99.5% (metals basis), powder -325 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11891.09 11891.22 11891.36
11891.09EA 36.5 EUR
11891.09 11891.22 11891.36
Cadmium ≥99.5% (metals basis), powder -325 mesh
Cadmium

Purity: 99.5% CAS No.: 7440-43-9 Appearance: Silver white UN No.: 3179 MDL No.: MFCD00010914

Formule: Cd
Kookpunt: 767 °C (1013 hPa)
Smeltpunt: 321 °C
Dichtheid: 8,64 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010914
CAS nummer: 7440-43-9
EINECS: 231-152-8
UN: 3179
ADR: 4.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR