Plunger dispensing cans
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10008 10108 10208 10308
119-0256EA 69.2 EUR
119-0256 119-0259 119-0263 119-0267
Plunger dispensing cans
Veiligheidscontainers
These plunger cans are used to dispense flammable liquids and solvents safely, without waste or exposure. The pump base is designed to leave less liquid in the bottom before having to refill. Perforated upper pan serves as a flame arrester and top rim has molded edge protector for smooth, trouble-free moistening.
  • Chemically resistant Ryton® and brass plunger
  • Galvanised steel body construction
Compliance with FM and TÜV approved.