Safety can for flammables, type I, nonmetallic
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
14160 14261 14561 14065Z 14160Z
119-0434EA 143 EUR
119-0434 119-0433 JUSU14561 119-0278 119-0279
Safety can for flammables, type I, nonmetallic
Veiligheidscontainers
Safety can constructed of high-density polyethylene to stand up to tough acids and corrosives, as well as heat.
  • Withstand tough acids, corrosives and heat
  • Dent, puncture and chip resistant
The self-closing cap, pressure-relief mechanism and flame arrester work together to prevent ignition sources from entering, without restricting liquid flow in or out of the can. All cans feature stainless steel hardware and flame arrester for superior chemical resistance. FM approved. Carbon Insert – A patented conductive current-carrying carbon insert is embeddedMore Product Information
Order Now
Specificaties
Environmentally Preferable