Essential Chromatography Pump Systems, C-900 PURE
Leverancier: BUCHI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11C90000 11C90010 11C90020 11C90030
11C90000.EA 6450 EUR
11C90000. 11C90010. 11C90020. 11C90030.
Essential Chromatography Pump Systems, C-900 PURE
Chromatografiesystemen Flash-chromatografiesystemen
C-900 PURE Essential chromatography pump system with flexible software control.
  • Easy accessibility and large touchscreen user interface
  • Basic design and setup
  • Intuitive handling
  • Small footprint and robust
  • Automated capabilities
  • Expansion possibilities
  • No limits for sample loading and fraction collection
The Pure Essential flash chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit fromMore Product Information