Calcium oxide 96-100.5% (excluding ignited substances), fine powder, Puriss., meets analytical specification of FCC, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12047-1KG 12047-5KG 12047-2.5KG
12047-1KGEA 37.2 EUR
12047-1KG 12047-5KG 12047-2.5KG
Calcium oxide 96-100.5% (excluding ignited substances), fine powder, Puriss., meets analytical specification of FCC, Fluka™
Calcium oxide
Formule: CaO
Kookpunt: 2,850 °C (1.013 hPa)
Dichtheid: 3,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010911
CAS nummer: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Order Now

Specification Test Results

Assay (excluding ignited substance) 96 - 100.5 %
Insoluble in HCl Max. 0.5 %
Loss on ignition Max. 3.5 %
Arsenic (As) Max. 0.0003 %
Lead (Pb) Max. 0.0002 %
Fluoride (F) Max. 0.015 %
Mg and alkali salts (SO₄) Max. 2.5 %
Form powder (fine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR