Platinum ≥99.98% (metals basis), powder -200 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12075.03 12075.06 12075.77
12075.03EA 286 EUR
12075.03 12075.06 12075.77
Platinum ≥99.98% (metals basis), powder -200 mesh
Platinum

Purity: 99.98% CAS No.: 7440-06-4 Odour: Odourless MDL No.: MFCD00011179

Formule: Pt
Kookpunt: 4530 °C (1013 hPa)
Smeltpunt: 1773 °C
Dichtheid: 21,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR