Calcium sulphate hemihydrate ≥97.0%, Purum, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12090-1KG 12090-25KG
12090-1KGEA 44.4 EUR
12090-1KG 12090-25KG
Calcium sulphate hemihydrate ≥97.0%, Purum, Fluka™
Calcium sulphate hemihydrate

  • ≥95% for general laboratory use.

Formule: CaSO₄·0.5H₂O
Smeltpunt: > 1400 °C
Dichtheid: 2,320 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00799067
CAS nummer: 10034-76-1
EINECS: 231-900-3

Order Now

Specification Test Results

Assay Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR