PIG® universal mats
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MAT203 MAT240 MAT412 MAT204 MAT231 MAT135 MAT139 MAT155 MAT216 MAT285 MAT251
121-0521EA 155 EUR
121-0521 121-0523 121-0534 121-0087 121-0094 121-0079 121-0081 121-0084 121-0089 NEWPMAT285 121-0047
PIG® universal mats
Absorptiematerialen en bescherming tegen morsen
These mats last 2 times longer than ordinary mats so it requires fewer change-outs. Save money with the mat that absorbs anything and works anywhere.
  • Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job
More Product Information