Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12104.09 12104.22 12104.36 12104.A3
12104.09EA 44.8 EUR
12104.09 12104.22 12104.36 12104.A3
Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade
Zirconium tetrachloride

Purity: 99.5+% CAS No.: 10026-11-6 Molecular Formula: ZrCl4 Formula Weight: 233.03 Physical Form: Powder Density: 2.803 Appearance: White to orange Melting Point: 331° subl. Solubility: Soluble in alcohol, ether. Decomposes in water forming ZrOCl2 and HCl Merck Reference: 14,10174 UN No.: 2503 MDL No.: MFCD00011306


Friedel-Crafts catalyst

Formule: Cl₄Zr
Kookpunt: 3577 °C (1013 hPa)
Dichtheid: 2,803
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011306
CAS nummer: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8,III
Merck Index: 14,10174

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR