Strontium chloride, anhydrous ≥95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12202.36 12202.A3
12202.36EA 79.8 EUR
12202.36 12202.A3
Strontium chloride, anhydrous ≥95%
Strontium chloride

Purity: 95% CAS No.: 10476-85-4 Molecular Formula: SrCl2 Formula Weight: 158.53 Physical Form: Powder Density: 3.052 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 873° Boiling Point: 1250° Merck Reference: 14,8840 MDL No.: MFCD00011249

Formule: Cl₂Sr
MW: 158.53 g/mol
Kookpunt: 1250 °C
Smeltpunt: 873 °C
Dichtheid: 3.052
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011249
CAS nummer: 10476-85-4
EINECS: 233-971-6
Merck Index: 14,8840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR