Erbium 99,9 REO basis, pieces, distilled dendritic

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Argon_filled

12255.14
12255.14EA 734 EUR
12255.14
Erbium 99,9 REO basis, pieces, distilled dendritic
Erbium

Purity: 99.9% CAS No.: 7440-52-0 Physical Form: Packaged under argon Sensitivity: Moisture Sensitive Appearance: Silver UN No.: 3178 MDL No.: MFCD00010987

Formule: Er
MW: 167.26 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010987
CAS nummer: 7440-52-0
EINECS: 231-160-1
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR