Vanadium(III) oxide ≥95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12301.30 12301.A1 12301.A7
12301.30EA 93.2 EUR
12301.30 12301.A1 12301.A7
Vanadium(III) oxide ≥95%
Vanadium(III) oxide

Purity: 95% CAS No.: 1314-34-7 Molecular Formula: O3V2 Formula Weight: 149.88 Physical Form: Powder Density: 4.87 Appearance: Black Odour: Odourless Melting Point: 1970° Solubility: Insoluble in water Merck Reference: 14,9923 UN No.: 3285 MDL No.: MFCD00011455

Formule: O₃V₂
Smeltpunt: 1970 °C
Dichtheid: 4,87
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011455
CAS nummer: 1314-34-7
EINECS: 215-230-9
UN: 3285
ADR: 6.1,II
Merck Index: 14,9923

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR