tetra-Chloroauric(III) acid hydrate (≥49% Au) ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

12325.03 12325.06 12325.14 12325.22 AB202976.0005
12325.03EA 189 EUR
12325.03 12325.06 12325.14 12325.22 ABCRAB202976.0005
tetra-Chloroauric(III) acid hydrate (≥49% Au) ≥99.9% (metals basis)
tetra-Chloroauric(III) acid hydrate

Purity: 99.9% CAS No.: 27988-77-8 Molecular Formula: HAuCl4.xH2O Formula Weight: 339.79(anhy) Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Yellow-orange UN No.: 3260 MDL No.: MFCD00149903

Formule: HAuCl₄·H₂O
MW: 357.8 g/mol
Dichtheid: 1.169 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149903
CAS nummer: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR