Iron(III) oxide ≥95%, purified, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12342-1KG 12342-250G 12342-6X1KG 12342-10KG
12342-1KGEA 64.6 EUR
12342-1KG 12342-250G 12342-6X1KG HONC12342-10KG
Iron(III) oxide ≥95%, purified, Fluka™
Iron(III) oxide
Formule: Fe₂O₃
Dichtheid: 5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011008
CAS nummer: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now

Specification Test Results

Assay Min. 95 %
Loss on ignition Max. 0.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR