Manganese(II) acetate tetrahydrate (∼22% Mn)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

12351.22 12351.36 12351.A3
12351.22EA 28.6 EUR
12351.22 12351.36 12351.A3
Manganese(II) acetate tetrahydrate (∼22% Mn)
Manganese(II) acetate tetrahydrate

CAS No.: 6156-78-1 Molecular Formula: C4H6MnO4.4H2O Formula Weight: 245.09 (173.03anhy) Physical Form: Crystalline Density: 1.589 Appearance: Pale pink Odour: Pungent Melting Point: 80° Solubility: Soluble in water, alcohol Merck Reference: 14,5724 MDL No.: MFCD00062552


Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst

Formule: (H₃CCOO)₂Mn·4H₂O
Smeltpunt: 80 °C
Dichtheid: 1,589
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00062552
CAS nummer: 6156-78-1
EINECS: 211-334-3
Merck Index: 14,5724

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR