Samarium 99,9 REO basis, foil, Thickness: 0.1 mm (0.004 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12421.FF 12421.FI
12421.FFEA 217 EUR
12421.FF 12421.FI
Samarium 99,9 REO basis, foil, Thickness: 0.1 mm (0.004 in)
Samarium

Purity: 99.9% CAS No.: 7440-19-9 Physical Form: ≈0.47g/25x25mm, Packaged in mylar under argon Odour: Odourless UN No.: 3178 MDL No.: MFCD00011233

Formule: Sm
MW: 150.36 g/mol
Kookpunt: 1900
Smeltpunt: 1076.85
Dichtheid: 7.42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011233
CAS nummer: 7440-19-9
EINECS: 231-128-7
UN: 3178
ADR: 4.1 II
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR