Tungsten(IV) carbide ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

12482.22 12482.36
12482.22EA 106 EUR
12482.22 12482.36
Tungsten(IV) carbide ≥99.5% (metals basis)
Tungsten(IV) carbide

Purity: 99.5% CAS No.: 12070-12-1 Molecular Formula: WC Formula Weight: 195.86 Physical Form: APS 1 micron Density: 15.63 Melting Point: 2870° Merck Reference: 14,9815 UN No.: 3178 MDL No.: MFCD00011464

Formule: CW
Kookpunt: 6000 °C (1013 hPa)
Smeltpunt: 2870 °C
Dichtheid: 15,63
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011464
CAS nummer: 12070-12-1
EINECS: 235-123-0
UN: 3178
ADR: 4.1,III
Merck Index: 14,9815

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR