Natriumdichromaat dihydraat ≥99%, Reagent Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Packaged under Argon in resealable ChemSeal™ bottles.

12611.22 12611.36 12611.A3
12611.22EA 28.2 EUR
12611.22 12611.36 12611.A3
Natriumdichromaat dihydraat ≥99%, Reagent Grade
Natriumdichromaat dihydraat

CAS No.: 7789-12-0 Molecular Formula: Cr2Na2O7.2H2O Formula Weight: 298.00 (261.97anhy) Physical Form: Crystalline Density: 2.348 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water Merck Reference: 14,8609 UN No.: 3087 MDL No.: MFCD00149166

Formule: Na₂Cr₂O₇·2H₂O
Smeltpunt: 100 °C (-2H₂O)
Dichtheid: 2,348
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149166
CAS nummer: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II
Merck Index: 14,8609

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR