Palladium(II) nitrate (10% Pd) (w/w) in aqueous solution

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12621.NB 12621.ND 12621.NH
12621.NBEA 590 EUR
12621.NB 12621.ND 12621.NH
Palladium(II) nitrate (10% Pd) (w/w) in aqueous solution
Palladium(II) nitrate

CAS No.: 10102-05-3 Molecular Formula: Pd(NO3)2 Formula Weight: 230.43 Physical Form: Liquid Appearance: Brown to yellow to orange UN No.: 3264 MDL No.: MFCD00011169

Formule: N₂O₆Pd
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011169
CAS nummer: 10102-05-3
EINECS: 233-265-8
UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR