Potassium chloride 99-100.5% (by argentometric titration), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E508 (max. 0.0001% Al), Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

12636-25KG 12636-1KG 12636-5KG 12636-6X1KG 12636-500G
12636-25KGEA 280 EUR
12636-25KG 12636-1KG 12636-5KG 12636-6X1KG 12636-500G
Potassium chloride 99-100.5% (by argentometric titration), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E508 (max. 0.0001% Al), Fluka™
Potassium chloride
Formule: KCl
Kookpunt: 1,413 °C (1.013 hPa)
Smeltpunt: 772 °C
Dichtheid: 2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (105°C, 3h) Max. 0.5 %
Acidity or alkalinity complying
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.5 ppm
Barium (Ba) complying
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 1 ppm
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Magnesium (Mg) Max. 0.005 %
Manganese (Mn) complying
Sodium (Na) Max. 0.07 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 5 ppm
Mg, earth alkali metals (as Ca) Max. 200 ppm
Bromide (Br) Max. 0.1 %
Iodide (I) test complying
Iodide (I) Max. 0.005 %
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR