Koper(II)sulfaat pentahydraat 99.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

12849-1KG 12849-25KG 12849-100G 12849-2.5KG 12849-500G 12849-6X1KG
12849-1KGEA 99.3 EUR
12849-1KG 12849-25KG 12849-100G 12849-2.5KG 12849-500G 12849-6X1KG
Koper(II)sulfaat pentahydraat 99.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
Koper(II)sulfaat pentahydraat
Formule: CuSO₄·5H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
ADR: 9,III
Bereik: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Assay (USP) 98.5 - 100.5 %
Loss on drying (250°C) 35.0 - 36.5 %
Arsenic (As) Max. 5 ppm
Calcium (Ca) Max. 50 ppm
Cobalt (Co) Max. 1 ppm
Iron (Fe) Max. 30 ppm
Potassium (K) Max. 100 ppm
Magnesium (Mg) Max. 50 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 20 ppm
Lead (Pb) Max. 4 ppm
Chloride (Cl) Max. 100 ppm
Substances not precipitated by H₂S (SO₄) Max. 0.3 %
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR