Titanium(II) hydride ≥99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

12857.06 12857.22 12857.36 12857.D3
12857.06EA 49 EUR
12857.06 12857.22 12857.36 12857.D3
Titanium(II) hydride ≥99% (metals basis)
Titanium(II) hydride

Purity: 99% CAS No.: 7704-98-5 Molecular Formula: H2Ti Formula Weight: 49.92 Physical Form: -325 Mesh Powder Density: 3.9 Appearance: Dark grey Odour: Odourless Melting Point: 450° dec. Merck Reference: 14,9473 UN No.: 1871 MDL No.: MFCD00151301

Formule: H₂Ti
Dichtheid: 3,9
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151301
CAS nummer: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,9473

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR