Mops, PowerHead™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PHU-124 PHU-154 PH6-154 PH6-150
129-0523EA 36.7 EUR
129-0523 129-0525 129-0521 MICRPH6-150
Mops, PowerHead™
Moppen
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
  • Stainless steel PowerHead™ handles hold and wring the various mop heads
  • Autoclavable
  • A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Bestelinformatie: All components are ordered separately.