Afvalzakken, Biohazard

Leverancier: STERLIN
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

509LHT 510LHT 510HT 511HT
129-0600EA 245 EUR
129-0600 129-0602 129-0601 129-0603
Afvalzakken, Biohazard
Zakken Afvalzakken
Sterilin autoclave bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilisation at 121 °C
  • Polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135 °C
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Convenient tissue box cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR